Rotary evaporators, ROdist digital
Toimittaja: Gardner Denver
ILMV112020EA 3750 EUR
ILMV112020 ILMV112033
Rotary evaporators, ROdist digital
This evaporator is equipped with a motorised height-adjustable lift with 'Safety Stop' function. If the power cuts off the evaporation flask automatically raises.
  • The evaporator features a compact design for minimal bench space
  • Space saving vertical glassware, coated (flexible design facilitates multiple glassware options)
  • Digital display with intuitive user interface
  • Free choice angle
  • Optional heating bath protection cover
  • Digital water-oil heating bath with integrated carrying handles
Tilaustiedot: Every ROdist digital is supplied complete with control drive, heating bath, vapor tube NS 29/32, seal, glassware (vertical, coated) with 1000 ml evaporation flask and 1000 ml receiving flask, clamps for glassware and woulf bottle. The Professional kit also includes the Ecoflex Speed Control System LVS 105 T-10 ef for faster processing and ultra quiet operation.