IP ActivFlo Biopsy II -kasetit
Toimittaja: Leica
720-2651EA 251 EUR
720-2651
IP ActivFlo Biopsy II -kasetit
Histologiset kasetit Kudos- ja biopsiakasetit
Surgipath ActivFlo Biopsy II cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lids have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Saatavana helposti tulostimeen ladattavissa teipatuissa pinoissa