Takki, puhdastila, BioClean-R™ Classic
Toimittaja: Nitritex
NITRCLC001-A10-44EA 0 EUR
NITRCLC001-A10-44 NITRCLC001-A10-52 NITRCLC001-A10-38 NITRCLC001-A10-46 NITRCLC001-A10-50 NITRCLC001-A10-40 NITRCLC001-A10-34 NITRCLC001-A10-48 NITRCLC001-A10-SP NITRCLC001-A10-42 NITRCLC001-A10-54 NITRCLC001-A10-36
Takki, puhdastila, BioClean-R™ Classic
These modern coats are made of MT811 fabric which provides protection for light duty tasks in non hazardous environments. The fabric MT811 is a twill weave fabric but its filtration performance at the smallest particle sizes is not quite as good as MT12 and MT031, which makes MT811 suitable for cleanroom classes ISO 7 and ISO 8.
  • Side fastening with polyester zip, stainless steel (nickel-free) slider
  • Mandarin collar, Raglan sleeves
  • Stud neck, stud wrists
Suitable for use in Class 10000, ISO 7, M5.5 environments.
Colour: White
Weight: 113 g/m²