Thermostatically controlled cabinets, TC series
Toimittaja: Lovibond Tintometer
132-0959EA 5950 EUR
132-0959 132-0957 132-0958 132-0955 132-0956 132-0954
Thermostatically controlled cabinets, TC series
Inkubaattorit BOD Incubators
The TC series of thermostatically controlled cabinets is used for continuous temperature control over a range of 2 to 40 °C. This makes them ideal for a wide range of different applications in industrial and research laboratories. In particular they are ideal for the temperature controlled storage of samples or BOD determination in effluent analysis work. Devices such as magnetic stirrers, which require a power supply, can be connected to sockets incorporated in the interior of the cabinet. Improved, robust, insulated housing and highly efficient components provide maximum energy efficiency.
  • Temperature range 2 to 40 °C, continuously adjustable in steps of 0,1 °C
  • Models with glass door: Insulating glass door in an ABS frame. Ceiling lighting, separately switchable
  • Models TC 135 S and TC 140 G are suitable for built under applications
  • Splash-proofed keypad, 2 buttons with tactile feedback
  • Integrated powerful cooling and heating. Coolant: R134a
There are four models available with standard doors, denoted by S, from 135 to 445 litres net capacity, and two models with glass doors, denoted by G, with 140 and 255 litres net capacity.
Sertifikaatit: The integral temperature control unit meets the requirements of the EMC directive issued as IEC 61326: 'Electrical devices for measurement, monitoring and for use in laboratories'.
Varoitus: * Dimensions are given with the work top (dimensions without work top: 600×600×819 mm)