Nylon film, Portex®
Toimittaja: Sims Portex
SIMS810032050200EA 117 EUR
SIMS810032300200 SIMS810032050200 SIMS810032200200
Nylon film, Portex®
Sulkijakalvot ja -matot Sulkijafilmit
Nylon film is ideal for sterilising and storing surgical instruments, dressings and linen. The permeability to ethylene oxide gas has been demonstrated by sterility tests on standard cultures, spore strips and chemical indicators placed inside sealed packets of film.