UV transilluminators
Toimittaja: Consort
CONSVLECX-15CEA 1790 EUR
CONSVLECX-15C CONSVLECX-15M CONSVLTFP-CWL
UV transilluminators
Transilluminaattorit UV-Transilluminaattorit
These transilluminators are based on an electronic ballast. The 25 kHz high frequency provides excellent stability and avoids any flickering effect.
  • Transilluminator heat is considerably reduced, avoiding the use of an extra cooling fan and thus reducing the noise
  • A fully adjustable safety screen, which can be positioned to suit the operator’s viewing angle, provides protection against exposure to harmful UV-rays
  • An intensity switch (except on the UV/white-light tables) allows you to select for both analysis (100%) and preparation (70%)
  • The lowest position enables you to visualise and cut fragments while minimising the risk of damaging the DNA
  • Unlimited filter life expectancy
  • The same model can be used from 100 to 240 VAC and from 50 to 60 Hz
  • Stainless steel frame equipped with an Ondulex® reflector