Teollisuusvaaka, suuren kapasiteetin vaa'at, IS -sarja
611-8623EA 695 EUR
VWRI611-8623 VWRI611-3573 VWRI611-8624 VWRI611-8622
Teollisuusvaaka, suuren kapasiteetin vaa'at, IS -sarja
A robust, reliable series of balances for a wide range of applications in the laboratory but also for general industrial use.
  • Backlit LCD with adjustable contrast
  • Rugged metal case with plastic, in-use protective cover
  • Selectable measuring units
  • Piece-counting, check-weighing, percentage weighing, animal weighing and density determination
  • Hook for below-balance weighing can be used for weighing large samples or solid density determination (software included)
  • Max. load determination (Peak hold)
Note: All these balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-8684) may be purchased for portable operation.
Lisävarusteiden tiedot: Optional Tablet System (611-8683) is suitable for use with models denoted with *.
Tilaustiedot: The balances can be equipped with optional Tablet system to expand functions and possibilities of weighing actions.
Toimitustiedot: Supplied with plastic in use protective cover.
Varoitus: Tablet Systems are not currently accredited for use with verified/EC-type approved balances.