Butterfly valve lockouts
Toimittaja: Brady
BRDY256960EA 113 EUR
BRDY256960
Butterfly valve lockouts
These lockouts were created to secure butterfly valves efficiently.
  • Non conductive
  • Allow two padlocks to be applied
Material: PP
Max. shackle diameter: 8 mm
Min. shackle clearance: 25 mm