Lisävarusteet, Storerooms for Target LoVial crimp top vials
Toimittaja: Thermo Scientific
NSCAC4075-1EA 1860 EUR
NSCAC4075-1
Lisävarusteet, Storerooms for Target LoVial crimp top vials
Koeputkitelineet Kromatografian näytekuppitelineet
Storerooms with target crimp-top vials offers a full size (nine-drawer) storage units complete with vials and caps.
  • Full size storeroom has 9 deep drawers with a capacity of 2000 vials and seals
  • Stackable cabinet constructed of heavy-gauge steel
  • With metal shelves and injection-molded drawers
  • Storeroom can be restocked as product is consumed
Toimitustiedot: Prestocked with 3000 each of: clear vials and preassembled clear crimp seals with FEP/red rubber septa