Racks for centrifuge tubes
Toimittaja: Greiner bio-one
GREI811250_5EA 51.5 EUR
GREI811250_5
Racks for centrifuge tubes
Koeputkitelineet Tube Racks
PP.
  • Autoclavable
  • Double platform
  • Easy to assemble
Colour: Grey/White