ddNTP set, 5 mM solutions (ddATP, ddCTP, ddGTP, ddTTP)
Toimittaja: Cytiva
27-2045-01EA 290 EUR
GEHE27-2045-01
ddNTP set, 5 mM solutions (ddATP, ddCTP, ddGTP, ddTTP)
Easy-to-use, time-saving solutions for use in any application requiring ddNTPs of high purity, such as dideoxy sequencing and genotyping reactions. Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase DNA Polymerase.
  • High purity: ≥98% triphosphate
  • Available as a convenient set containing ddATP, ddCTP, ddGTP, and ddTTP in separate tubes
  • Supplied in purified water as 5 mM solutions