4-Hydroxyphenyl glyoxal (HPG)
Toimittaja: G Bioscienes
Synonyms: HPG
BC94EA 497 EUR
BC94
4-Hydroxyphenyl glyoxal (HPG)
4-Hydroxyphenyl glyoxal (HPG)
G-Biosciences' HPG (p-hydroxyphenylglyoxal) specifically reacts with arginine residues under mild conditions (pH 7 to 9, 25°C) to yield spectrophotometrically measurable signal for amino acid detection.
G-Biosciences' HPG is more water-soluble and more resistant to oxidation than other arginine modifiers. Modification can be monitored at 340 nm.
Kaava: C₈H₆O₃
MW: 150,131 g/mol
MDL numero: MFCD00054984
CAS-numero: 24645-80-5
https://fi.vwr.com/stibo/web/std.lang.all/44/25/23914425.jpg