Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)
Toimittaja: G Bioscienes
Synonyms: Sulfo NHS
BC97EA 461 EUR
BC97
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)
Sulpho-NHS is water-soluble, long-lived, and hydrolyses relatively slowly in water and in aqueous buffer, unlike NHS.
Kaava: C₄H₄NNaO₆S
MW: 217,13 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00043100
CAS-numero: 106627-54-7
https://fi.vwr.com/stibo/web/std.lang.all/44/54/23914454.jpg