The iMini Plus PDF loggers are designed for the monitoring of temperature sensitive shipments to distant destinations, where proprietary software and interfaces are unavailable. Report generation includes a summary of transport condition, time spent out of specification and trip statistics in addition to date, time and temperature of all readings plus a graph of the temperature of the trip. The PDF report can be validated by Console software by emailing the PDF report and binary files to the supplier thus meeting the requirements for 21 CFR Part11.
- Large LCD display with multi function viewing
- Advanced multiple alarm settings
- Bookmarking and continuous logging features
- PDF report can be customised to display graph, list of readings and summary of alarm limits violations
- Simple USB to USB cable for PC connection and travels with the logger in the consignment
- Restart feature by START button
- Case material: Polycarbonate/ABS
Alarm thresholds is two and alarm configuration is high or low. User Programmable, auto restart and stop option is available.
LCD operating range: −20 to +70 °C
Sertifikaatit: ISO 9001:2008, RoHS, EN 12830, IP 65 (with palsticcap over USB connecter), NIST traceable upon request.
Toimitustiedot: Supplied with cable and battery. 24 months warranty, excluding battery.