Kalsiumvetyfosfaatti, vedetön ≥98.0%, jauhe, BAKER ANALYZED®, J.T.Baker®

1430-09
Kalsiumvetyfosfaatti, vedetön ≥98.0%, jauhe, BAKER ANALYZED®, J.T.Baker®
Kalsiumvetyfosfaatti
Kaava: CaHPO₄
MW: 136,06 g/mol
Sulamispiste: 370 °C
Tiheys: 2,89 g/cm³ (20 °C)
MDL numero: MFCD00010909
CAS-numero: 7757-93-9
EINECS: 231-826-1
Merck Index: 12,01739

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use
Assay (CaHPO₄) ≥98.0 %
Chloride (Cl) ≤0.25 %
Sulfate (SO₄) ≤0.05 %
Nitrate (NO₃) ≤0.005 %
Arsenic (As) ≤3 ppm
Heavy Metals (as Pb) ≤0.003 %
Iron (Fe) ≤0.005 %
Insoluble in HCl ≤0.09 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR