VWR® Signature™, Pipetinkärkiä
Toimittaja: VWR
VWR® Signature™
613-2442EA 118 EUR
613-2442
VWR® Signature™, Pipetinkärkiä
Pipetinkärjet
General purpose tips made from PP for use with Sartorius® Proline pipettors.
  • Sustainable design based on virgin resins
  • Tips are Rnase/Dnase-, endotoxin- and DNA-free
General purpose tips made from PP for use with Sartorius® Proline pipettors.
Up to 250 µl headspace prevents overaspiration pipettor contamination.