deconex® 11 UNIVERSAL
Mildly alkaline cleaning liquid concentrate for laboratory glassware. Suitable for soaking baths and ultrasonic cleaning equipment.
With active chlorine
Dosage 1 - 2% for normal cleaning; 2 - 5% for intensive cleaning at 20 - 30 °C"
deconex® 12 BASIC
Mildly alkaline cleaning liquid concentrate for laboratory glassware. Suitable for soaking baths and ultrasonic cleaning equipment.
pH 1% solution: 11,8
Dosage 1 - 2% for normal cleaning; 2 - 5% for intensive cleaning at 20 - 30 °C
Not suitable for aluminium, tin, zinc"
deconex® 12 PA-x
High performance cleaning concentrate for use in research, medicine, biology and chemistry as well as for optical and electronic components. For soaking baths and ultrasonic cleaning systems.
NTA free
Dosage 0,5 - 1% for normal cleaning; 2 - 4% for intensive cleaning at 20 - 30 °C
deconex® 15 NF. Alkaline cleaning liquid concentrate for laboratory glassware. Particularly suitable for soaking baths and ultrasonic cleaning equipment as a pretreatment for washing machine cleaning.
Phosphate-free, non foaming
Dosage 2 - 3% for normal cleaning; 3 - 5% for intensive cleaning at 20 - 30 °C
deconex® 15 PF-x. Alkaline cleaning liquid concentrate for laboratory glassware. Suitable for soaking baths and ultrasonic cleaning equipment where phosphate levels are critical in the application.
Phosphate-free
Dosage 1 - 2% for normal cleaning; 2 - 5% for intensive cleaning at 20 - 30 °C
deconex® 20 NS-x. High performance alkaline cleaning concentrate for laboratory equipments in research, medicine, biology and chemistry as well as for optical and electronic components. For use in soaking baths, ultrasonic cleaning systems and cleaning in washing machines.
pH 1% solution: 11,9
Dosage 2 - 5% for normal cleaning; 5 - 10% for intensive cleaning at 20 - 30 °C
Not suitable for aluminium, tin, zinc, non ferrous metals
Varoitus: Note: Most of the materials are corrosive or irritant and care must be taken with with choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.