These ion selective electrodes feature a replaceable gel reference electrolyte cartridge and a fixed cable with BNC connector. Compatible with Hach sensION, sensION+ and H-Series systems.
- Replaceable gel electrolyte cartridge: Maximum life through reduced maintenance
- Provide fast, stable, and accurate results
- Easy to use
- Analysis packages contain everything that is need for ISE measurements in one package
Nitrate analysis packages includes: sensION combination Platinum Series Nitrate ISE, 6-pack of replacement nitrate membrane modules, two replacement electrolyte cartridges, internal filling solution (50 ml), two Hach nitrate standard solutions (10 mg/L (500 ml) and 100 mg/L (500 ml)), Hach nitrate ionic strength adjustor (ISA) Powder Pillows (pack of 100), 50-ml beaker and ISE User Manual.
Toimitustiedot: Electrodes are supplied with two replacement electrolyte cartridges and user manual.