Eye showers, free-standing
Toimittaja: asecos
MSGA18051EA 500 EUR
MSGA18051
Eye showers, free-standing
These free-standing eye showers feature two shower heads with a large flushing area and a plastic basin.
  • Shower heads covered with rubber protection and dust caps
  • Dust-protection-cap easily falls off during use
  • Large pressure plate, easy to use
  • Large distance between the shower heads for efficient cleaning of eyes
Water connection: ¹/₂"
Water flow rate 12 L/min
Min. operating pressure: 1,5 to 2 bar.
Sertifikaatit: ISO3864-1