Foil Cover-ALLS, Bel-Art ProCulture, SP Bel-Art
Toimittaja: Bel-Art
SP Bel-Art ProCulture
BELAF132001035EA 87.2 EUR
BELAF132001035
Foil Cover-ALLS, Bel-Art ProCulture, SP Bel-Art
Pullon suojat, alumiinia
Used in cell culture to cover flasks while allowing proper oxygenation, these economical and disposable covers also provide dust-free storage of clean labware.
  • Provides a temporary, sterile boundary for autoclave sterilization of labware
  • Preformed to quickly cover a wide variety of labware using only one hand
Great for use in centralized washing areas where cleaned labware needs to be transported back to the lab while remaining clean and contaminant-free.