Test tubes
Toimittaja: Greiner bio-one
CELLSTAR®
211-3116EA 278 EUR
211-3116
Test tubes
Putket Koeputket
Universal tubes, made of PP, translucent, for a wide range of applications.
  • Graduated
  • With two-position vent stopper
  • Sterile