Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 8C12]

Toimittaja: AbFrontier
ABFRLF-MA0107EA 267 EUR
ABFRLF-MA0107
Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 8C12]
Vasta-aineet
Antithrombin III is a plasma protein synthesized in the liver and has molecular weight of 58kDa. Antithrombin III is a heparin-binding glycoprotein that acts as the major inhibitor of thrombin. Also it interferes with several plasma proteases, including kallikrein and factors IXa, Xa, XIa and XIIa, thereby playing a central role in regulating coagulation. It is also used as a therapeutic agent in patients with sepsis and disseminated intravascular coagulation. In addition, a number of recent studies have shown that antithrombin III has anti-inflammatory properties, which are independent of its effects on coagulation.

Western Blot: 1:1000;

Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: Monoclonal
Clone: 8C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR