Tubulin is a heterodimer consisting of α and β-tubulin subunits. It is nearly ubiquitous among eukaryotic cells. The α and β tubulins, which are each about 55 kDa MW, are homologous but not identical. α-β tubulin heterodimer is the basic building block of microtubules. This intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis
Western Blot: 1:10,000;
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat