Anti-CALCA Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAM2021AEA 348 EUR
ABGEAM2021A
Anti-CALCA Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CALCA (calcitonin-related polypeptide alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR