Anti-OR52I2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP11831BEA 348 EUR
ABGEAP11831B
Anti-OR52I2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: OR52I2 (olfactory receptor, family 52, subfamily I, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR