Anti-CDH24 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP12160AEA 385 EUR
ABGEAP12160A
Anti-CDH24 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CDH24 (cadherin 24, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR