Anti-PDCD1LG2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP12309AEA 348 EUR
ABGEAP12309A
Anti-PDCD1LG2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PDCD1LG2 (programmed cell death 1 ligand 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR