Anti-TTLL4 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP12581CEA 348 EUR
ABGEAP12581C
Anti-TTLL4 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TTLL4 (tubulin tyrosine ligase-like family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR