Anti-ADAMTS5 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP13847AEA 348 EUR
ABGEAP13847A
Anti-ADAMTS5 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ADAMTS5 (ADAM metallopeptidase with thrombospondin type 1 motif, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR