Anti-ERN1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP14250BEA 348 EUR
ABGEAP14250B
Anti-ERN1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ERN1 (endoplasmic reticulum to nucleus signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR