Anti-MEOX1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP14566CEA 385 EUR
ABGEAP14566C
Anti-MEOX1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MEOX1 (mesenchyme homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR