Anti-USP8 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP2138BEA 348 EUR
ABGEAP2138B
Anti-USP8 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: USP8 (ubiquitin specific peptidase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR