Anti-CALCR Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP5688BEA 385 EUR
ABGEAP5688B
Anti-CALCR Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CALCR (calcitonin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR