Anti-HIF1A Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP7759CEA 348 EUR
ABGEAP7759C
Anti-HIF1A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR