Anti-HCLS1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2329AEA 350 EUR
ABGEAT2329A
Anti-HCLS1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HCLS1 (hematopoietic cell-specific Lyn substrate 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human