Anti-HIP1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2370AEA 350 EUR
ABGEAT2370A
Anti-HIP1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human