Anti-HIPK1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2373AEA 350 EUR
ABGEAT2373A
Anti-HIPK1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HIPK1 (homeodomain interacting protein kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human