Anti-HMGB1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2385AEA 350 EUR
ABGEAT2385A
Anti-HMGB1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human