Anti-HPRT1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2434AEA 350 EUR
ABGEAT2434A
Anti-HPRT1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HPRT1 (hypoxanthine phosphoribosyltransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human