Anti-HSPA1B Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2444AEA 350 EUR
ABGEAT2444A
Anti-HSPA1B Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HSPA1B (heat shock 70kDa protein 1B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human