Anti-ICA1 Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT2467AEA 350 EUR
ABGEAT2467A
Anti-ICA1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ICA1 (islet cell autoantigen 1, 69kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR