Anti-IL13RA1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2503AEA 350 EUR
ABGEAT2503A
Anti-IL13RA1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IL13RA1 (interleukin 13 receptor, alpha 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human