Anti-IRF1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2540AEA 350 EUR
ABGEAT2540A
Anti-IRF1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IRF1 (interferon regulatory factor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human