Anti-IRF4 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2545AEA 350 EUR
ABGEAT2545A
Anti-IRF4 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IRF4 (interferon regulatory factor 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human