Anti-ISG20 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2557AEA 350 EUR
ABGEAT2557A
Anti-ISG20 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ISG20 (interferon stimulated exonuclease gene 20kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human