Anti-MAPK13 Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT2788AEA 350 EUR
ABGEAT2788A
Anti-MAPK13 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MAPK13 (mitogen-activated protein kinase 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR