Anti-MBNL1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2813AEA 350 EUR
ABGEAT2813A
Anti-MBNL1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MBNL1 (muscleblind-like splicing regulator 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human