Anti-MEF2D Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2838AEA 350 EUR
ABGEAT2838A
Anti-MEF2D Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MEF2D (myocyte enhancer factor 2D)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human