Anti-MFNG Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2853AEA 350 EUR
ABGEAT2853A
Anti-MFNG Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MFNG (MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human