Anti-MIF Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT2870AEA 350 EUR
ABGEAT2870A
Anti-MIF Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR